missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-(1-Piperidinyl)pyridine-3-carboxaldehyde, 98%

Product Code. 11494580
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494580 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11494580 Supplier Alfa Aesar™ Supplier No. H50127.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 241816-11-5
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
MDL Number MFCD04117814
InChI Key RKIMIISNVCLRLA-UHFFFAOYSA-N
Synonym 6-piperidin-1-ylnicotinaldehyde, 6-piperidin-1-yl nicotinaldehyde, 6-piperidin-1-yl pyridine-3-carbaldehyde, 6-piperidinonicotinaldehyde, 6-1-piperidinyl nicotinaldehyde, 6-1-piperidinyl pyridine-3-carboxaldehyde, 6-piperidylpyridine-3-carbaldehyde, acmc-20alxp, 3-pyridinecarboxaldehyde,6-1-piperidinyl, 3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-5'-carbaldehyde
PubChem CID 2763980
IUPAC Name 6-piperidin-1-ylpyridine-3-carbaldehyde
SMILES C1CCN(CC1)C2=NC=C(C=C2)C=O

Specifications

Melting Point 42°C to 44°C
Quantity 1 g
Formula Weight 190.25
Percent Purity 98%
Chemical Name or Material 6-(1-Piperidinyl)pyridine-3-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.