missing translation for 'onlineSavingsMsg'
Learn More

6-(1-Pyrrolidinyl)nicotinaldehyde, Thermo Scientific™

Product Code. 10360643
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
Product Code. Quantity Packaging unitSize
10360643 1 g Amber Glass Bottle 1g
10299392 250 mg Amber Glass Bottle 250mg
10316804 5 g Amber Glass Bottle 5g
3 options
This item is not returnable. View return policy

Product Code. 10360643

Brand: Thermo Scientific Maybridge MO07079DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 261715-39-3
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.219
InChI Key FFFAXGTZBUMKTI-UHFFFAOYSA-N
Synonym 6-1-pyrrolidinyl nicotinaldehyde, 6-pyrrolidin-1-yl nicotinaldehyde, 6-pyrrolidin-1-yl pyridine-3-carbaldehyde, 6-pyrrolidin-1-yl pyridine-3-carboxaldehyde, 6-1-pyrrolidinyl pyridine-3-carboxaldehyde, 6-pyrrolidin-1-ylnicotinaldehyde, 6-pyrrolidinylpyridine-3-carbaldehyde, 6-1-pyrrolidino pyridine-3-carboxaldehyde, acmc-1cg5q, 6-1-pyrrolidino nicotinaldehyde
PubChem CID 2794775
IUPAC Name 6-pyrrolidin-1-ylpyridine-3-carbaldehyde
SMILES C1CCN(C1)C2=NC=C(C=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 176.22
Percent Purity 97%
Chemical Name or Material 6-(1-Pyrrolidinyl)nicotinaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.