missing translation for 'onlineSavingsMsg'
Learn More

6-(2-Furyl)nicotinaldehyde, 97%, Thermo Scientific™

Product Code. 10285614
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10285614 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10285614 Supplier Thermo Scientific Maybridge Supplier No. CC54904DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 886851-42-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H7NO2
MDL Number MFCD04115423
Quantity 1 g
Synonym 6-2-furyl nicotinaldehyde, 6-furan-2-yl pyridine-3-carbaldehyde, 6-furan-2-yl nicotinaldehyde, 6-2-furyl pyridine-3-carbaldehyde, 6-2-furanyl-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde,6-2-furanyl, 6-furan-2-yl-pyridine-3-carbaldehyde
InChI Key RHHAYCIYWCFECO-UHFFFAOYSA-N
SMILES C1=COC(=C1)C2=NC=C(C=C2)C=O
IUPAC Name 6-(furan-2-yl)pyridine-3-carbaldehyde
Molecular Weight (g/mol) 173.171
PubChem CID 24729675
Formula Weight 173.17
Percent Purity 97%
Chemical Name or Material 6-(2-Furyl)nicotinaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.