missing translation for 'onlineSavingsMsg'
Learn More

6-(2-furyl)nicotinic acid, Thermo Scientific™

Product Code. 10557484
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10557484 5 g 5g
10001934 250 mg 250mg
10144913 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10557484 Supplier Thermo Scientific Maybridge Supplier No. CC54901DE
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-36-2
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD06410385
InChI Key ALJRNVXLRKHVQF-UHFFFAOYSA-N
Synonym 6-2-furyl nicotinic acid, 6-furan-2-yl nicotinic acid, 6-furan-2-yl pyridine-3-carboxylic acid, 6-2-furyl pyridine-3-carboxylic acid
PubChem CID 19010290
IUPAC Name 6-(furan-2-yl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=C(C=C1)C1=CC=CO1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 6-(2-Furyl)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.