missing translation for 'onlineSavingsMsg'
Learn More

6-(4-Bromophenyl)-4,5-dihydro-3(2H)-pyridazinone, 98%, Thermo Scientific™

Product Code. 11430070
Change view
Click to view available options
Quantity:
1 g
5 g
missing translation for 'unitSize':
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
11430070 1 g 1g
11440070 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11430070 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' H50588.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36725-37-8
Molecular Formula C10H9BrN2O
Molecular Weight (g/mol) 253.099
MDL Number MFCD00203490
InChI Key CDDNLMGOLYMTSF-UHFFFAOYSA-N
Synonym 6-4-bromophenyl-4,5-dihydropyridazin-3 2h-one, 6-4-bromophenyl-4,5-dihydro-2h-pyridazin-3-one, 6-4-bromophenyl-2,3,4,5-tetrahydropyridazin-3-one, 6-4-bromophenyl-4,5-dihydro-3 2h-pyridazinone, 6-4-bromophenyl-2,4,5-trihydropyridazin-3-one, 3-4-bromophenyl-4,5-dihydro-1h-pyridazin-6-one, 6-4-bromophenyl-4 5-dihydro-2h-pyrida, 6-p-bromophenyl-4,5-dihydro-3 2h-pyridazinone, 3 2h-pyridazinone,6-4-bromophenyl-4,5-dihydro, 6-4-bromo-phenyl-4,5-dihydro-2h-pyridazin-3-one
PubChem CID 689108
IUPAC Name 3-(4-bromophenyl)-4,5-dihydro-1H-pyridazin-6-one
SMILES C1CC(=O)NN=C1C2=CC=C(C=C2)Br

Specifications

Melting Point 167°C to 171°C
Quantity 1 g
Formula Weight 253.1
Percent Purity 98%
Chemical Name or Material 6-(4-Bromophenyl)-4,5-dihydro-3(2H)-pyridazinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.