missing translation for 'onlineSavingsMsg'
Learn More

6-[4-(tert-Butoxycarbonyl)piperazin-1-yl]nicotinic acid, ≥97%, Thermo Scientific™

Product Code. 10639954
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10639954 1 g Amber Glass Bottle 1g
10219442 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10639954 Supplier Thermo Scientific Maybridge Supplier No. CC43901DA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 201809-22-5
Molecular Formula C15H21N3O4
Molecular Weight (g/mol) 307.35
MDL Number MFCD06245543
InChI Key PWYGTZUOLAGDNK-UHFFFAOYSA-N
Synonym 6-4-tert-butoxycarbonyl piperazin-1-yl nicotinic acid, 6-4-tert-butoxycarbonyl piperazin-1-yl pyridine-3-carboxylic acid, 4-5-carboxy-pyridin-2-yl-piperazine-1-carboxylic acid tert-butyl ester, 6-4-tert-butoxy carbonyl piperazin-1-yl pyridine-3-carboxylic acid, 6-4-2-methylpropan-2-yl oxycarbonyl piperazin-1-yl pyridine-3-carboxylic acid, n4-boc-6-piperazin-1-ylnicotinic acid, 6-4-tert-butoxycarbonyl piperazino nicotinic acid, 6-4-tert-butoxycarbonyl piperazino-nicotinic acid, 1-piperazinecarboxylicacid,4-5-carboxy-2-pyridinyl-,1-1,1-dimethylethyl ester
PubChem CID 2771823
IUPAC Name 6-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]pyridine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=C(C=C1)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 307.35
Percent Purity 97%
Chemical Name or Material 6-[4-(Tert-butoxycarbonyl)piperazin-1-yl]nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.