missing translation for 'onlineSavingsMsg'
Learn More

6,6'-Methylenebis(2-(tert-butyl)-4-ethylphenol), TRC

Product Code. 30468604
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 g
missing translation for 'unitSize':
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30468604 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30468604 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM3044482.5G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6,6'-Methylenebis(2-(tert-butyl)-4-ethylphenol)
CAS 88-24-4
Molecular Formula C25 H36 O2
Quantity 2.5 g
Synonym Phenol','2,2'-methylenebis[6-tert-butyl-4-ethyl- (6CI,7CI,8CI), 2,2'-Methylenebis[6-(1,1-dimethylethyl)-4-ethylphenol], 2,2-Methylenebis(4-ethyl-6-t-butylphenol), 2,2-Methylenebis(4-ethyl-6-tert-butylphenol), 2,2'-Methylenebis(6-tert-butyl-4-ethylphenol), 2,2'-Methylenebis[4-ethyl-6-tert-butylphenol], 2,2'-Methylidene bis-4-ethyl-6-tert-butylphenol), AO 425, AW 500, Agidol 7, Antage W 500, Antioxidant 425, Bis(2-hydroxy-3-tert-butyl-5-ethylphenyl)methane, Bis(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methane, Chemanox 22, Cyanox 425, MEB, NS 5, NSC 7782, Nocrac NS 5, Nonflex EBP, Plastanox 425, Swanox 425, Yoshinox 425
InChI Formula InChI=1S/C25H36O2/c1-9-16-11-18(22(26)20(13-16)24(3,4)5)15-19-12-17(10-2)14-21(23(19)27)25(6,7)8/h11-14,26-27H,9-10,15H2,1-8H3
SMILES CCc1cc(Cc2cc(CC)cc(c2O)C(C)(C)C)c(O)c(c1)C(C)(C)C
IUPAC Name 2-tert-butyl-6-[(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methyl]-4-ethylphenol
Molecular Weight (g/mol) 368.55
Formula Weight 368.2715
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.