missing translation for 'onlineSavingsMsg'
Learn More

(6,7-Dihydro-4H-thieno[3,2-c]pyran-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10133824
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10133824 250 mg Amber Glass Bottle 250mg
10193864 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10133824

Brand: Thermo Scientific Maybridge CC61114CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 63932-26-3
Molecular Formula C8H11NOS
Molecular Weight (g/mol) 169.242
MDL Number MFCD08690319
InChI Key NMVKUIFBCHQTHO-UHFFFAOYSA-N
Synonym 4h,6h,7h-thieno 3,2-c pyran-4-ylmethanamine, 6,7-dihydro-4h-thieno 3,2-c pyran-4-yl methylamine, 4h-thieno 3,2-c pyran-4-methanamine,6,7-dihydro, 6,7-dihydro-4h-thieno-3,2-c pyran-4-ylmethyl amine, 6,7-dihydro-4h-thieno 3,2-c pyran-4-yl methanamine, 1-4h,6h,7h-thieno 3,2-c pyran-4-yl methanamine, 4h,6h,7h-thieno 3,2-c pyran-4-yl methanamine, 6,7-dihydro-4h-thiopheno 3,2-c pyran-4-ylmethylamine, 1-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl methanamine
PubChem CID 12225896
IUPAC Name 6,7-dihydro-4H-thieno[3,2-c]pyran-4-ylmethanamine
SMILES C1COC(C2=C1SC=C2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 169.25
Percent Purity 97%
Chemical Name or Material (6, 7-Dihydro-4H-thieno[3, 2-c]pyran-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.