missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydro-5H-pyrrolo[3,4-b]pyridine dihydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 15234794 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15234794 1 g Glass bottle 1g
15224794 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15234794 Supplier Thermo Scientific Acros Supplier No. 449900010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147740-02-1
Molecular Formula C7H8N2·2ClH
Molecular Weight (g/mol) 193.08
InChI Key FZBCVKVGLQRBHY-UHFFFAOYSA-N
Synonym 6,7-dihydro-5h-pyrrolo 3,4-b pyridine dihydrochloride, 5h,6h,7h-pyrrolo 3,4-b pyridine dihydrochloride, 6,7-dihydro-5h-pyrrolo 3,4-b pyridine 2hcl, 5h-pyrrolo 3,4-b pyridine, 6,7-dihydro-, dihydrochloride, pubchem18111, acmc-1c7fx, 6,7-dihydro-5h-pyrrolo 3,4-b pyridine dihcl, 6,7-dihydro-5h-pyrrolo 3,4-b pyridine dihydrochlor, 5,7-dihydro-6h-pyrrolo 3,4-b pyridine dihydrochloride
PubChem CID 23503616
IUPAC Name 6,7-dihydro-5H-pyrrolo[3,4-b]pyridine;dihydrochloride
SMILES C1C2=C(CN1)N=CC=C2.Cl.Cl

Specifications

Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
Formula Weight 193.08
Percent Purity 97%
Chemical Name or Material 6,7-Dihydro-5H-pyrrolo[3,4-b]pyridine dihydrochloride
compliance-icons
Product Identifier
  • 6,7-Dihydro-5H-pyrrolo¢3,4-b!pyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.