missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydrobenzo[b]thiophen-4-one, 98%

Product Code. 15456698
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. Quantity unitSize
15456698 10 g 10g
15446698 2 g 2g
2 options
This item is not returnable. View return policy

Product Code. 15456698

Brand: Thermo Scientific Alfa Aesar H60220.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13414-95-4 | C8H8OS | 152.211 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

Solubility
Soluble in chloroform.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13414-95-4
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.211
MDL Number MFCD00005861
InChI Key GJEKNELSXNSYAQ-UHFFFAOYSA-N
Synonym 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro, 6,7-dihydro-1-benzothiophen-4 5h-one, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 4,5,6,7-tetrahydro-4-benzothiophenone
PubChem CID 83418
IUPAC Name 6,7-dihydro-5H-1-benzothiophen-4-one
SMILES C1CC2=C(C=CS2)C(=O)C1

Specifications

Melting Point 38°C to 40°C
Boiling Point 120°C
Flash Point 110°C (230°F)
Refractive Index 1.5895
Quantity 10 g
Beilstein 112276
Solubility Information Soluble in chloroform.
Formula Weight 152.21
Percent Purity 98%
Chemical Name or Material 6,7-Dihydrobenzo[b]thiophen-4-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.