missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydroxy-3,4-dihydroisoquinoline, 98%, Thermo Scientific™

Product Code. 10257550 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10257550

Brand: Thermo Scientific Acros 277830010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4602-83-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 244.0°C to 248.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H9NO2
MDL Number MFCD00143490
Quantity 1 g
Synonym 3,4-dihydroisoquinoline-6,7-diol, 6,7-dihydroxy-3,4-dihydroisoquinoline, 6,7-isoquinolinediol, 3,4-dihydro
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in hot methanol,soluble in hot chloroform-methanol (1:1)
InChI Key WQFVTSPRATYMNQ-UHFFFAOYSA-N
SMILES C1CNC=C2C1=CC(=O)C(=C2)O
IUPAC Name 7-hydroxy-3,4-dihydro-2H-isoquinolin-6-one
Molecular Weight (g/mol) 163.18
PubChem CID 5922170
Formula Weight 163.18
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 6, 7-Dihydroxy-3, 4-dihydroisoquinoline, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 6,7-Dihydroxy-3,4-dihydroisoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.