missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%

Product Code. 11480637
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11480637 5 g 5g
11490637 25 g 25g
11400647 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11480637

Brand: Thermo Scientific Alfa Aesar B22469.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2328-12-3 | C11H16ClNO2 | 229.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2328-12-3
Molecular Formula C11H16ClNO2
Molecular Weight (g/mol) 229.70
MDL Number MFCD00012744
InChI Key SHOWAGCIRTUYNA-UHFFFAOYSA-N

Specifications

Melting Point 256°C to 260°C
Quantity 5 g
Beilstein 3634126
Synonym 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, heliamine hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolinehydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 1,2,3,4-tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride, isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-, hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline, chloride, pubchem22149, acmc-1cjku, ksc203s7f
SMILES COC1=C(C=C2CNCCC2=C1)OC.Cl
IUPAC Name 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
PubChem CID 6456035
Formula Weight 229.71
Percent Purity 97%
Chemical Name or Material 6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.