missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific™

Product Code. 10507980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10507980 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10507980 Supplier Thermo Scientific Acros Supplier No. 292980050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 2328-12-3
Molecular Formula C11H15NO2·ClH
Molecular Weight (g/mol) 229.71
InChI Key SHOWAGCIRTUYNA-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, heliamine hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolinehydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 1,2,3,4-tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride, isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-, hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline, chloride, pubchem22149, acmc-1cjku, ksc203s7f
PubChem CID 6456035
IUPAC Name 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES COC1=C(C=C2CNCCC2=C1)OC.Cl

Especificaciones

CAS Min % 97.5
CAS Max % 100.0
Melting Point 252.0°C to 254.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 229.71
Percent Purity 98%
Physical Form Shiny Crystalline Powder or Flakes
Chemical Name or Material 6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.