missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxyquinazoline-2,4-dione, 98%

Product Code. 11368145
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368145 10 g 10g
11378145 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368145 Supplier Thermo Scientific Alfa Aesar Supplier No. L08411.09
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 28888-44-0 | C10H10N2O4 | 222.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28888-44-0
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.20
MDL Number MFCD00023889
InChI Key KWNQIIMVPSMYEM-UHFFFAOYSA-N
Synonym 6,7-dimethoxyquinazoline-2,4-dione, 2,4-dihydroxy-6,7-dimethoxyquinazoline, 6,7-dimethoxyquinazoline-2,4 1h,3h-dione, 6,7-dimethoxyquinazoline-2,4-diol, 6,7-dimethoxy-1,2,3,4-tetrahydroquinazoline-2,4-dione, 6,7-dimethoxy-1h,3h-2,4-quinazolindione, 2,4 1h,3h-quinazolinedione, 6,7-dimethoxy, 6,7-dimethoxy-2,4-quinazolinedione, 2,4-dihydroxy-6,7-dimethoxy quinazoline, 6,7-dimethoxy-2,4-dihydroxy quinazoline
PubChem CID 120081
IUPAC Name 6,7-dimethoxy-1H-quinazoline-2,4-dione
SMILES COC1=C(OC)C=C2C(=O)NC(=O)NC2=C1

Specifications

Melting Point >300°C
Quantity 10 g
Beilstein 204597
Formula Weight 222.2
Percent Purity 98%
Chemical Name or Material 6,7-Dimethoxyquinazoline-2,4-dione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.