missing translation for 'onlineSavingsMsg'
Learn More

6,8,11-Trihydroxy-1-methoxy-5,12-naphthacenedione (Doxorubicin Impurity), 6,8,11-Trihydroxy-1-methoxy-5,12-naphthacenedione (Doxorubicin Impurity), TRC

Product Code. 30008845
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008845 10 mg 10mg
30009353 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008845 Supplier Toronto Research Chemicals Supplier No. TRCT79521010MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C19H12O6
Molecular Weight (g/mol) 336.3
Synonym 7,8-Dehydro-9,10-Desacetyl Doxorubicinone
IUPAC Name 6,8,11-trihydroxy-1-methoxytetracene-5,12-dione
SMILES COc1cccc2C(=O)c3c(O)c4cc(O)ccc4c(O)c3C(=O)c12

Specifications

CAS 64845-67-6
Quantity 10 mg
InChI Formula InChI=1S/C19H12O6/c1-25-12-4-2-3-10-13(12)19(24)15-14(17(10)22)18(23)11-7-8(20)5-6-9(11)16(15)21/h2-7,20-21,23H,1H3
Formula Weight 336.0634
Chemical Name or Material 6,8,11-Trihydroxy-1-methoxy-5,12-naphthacenedione (Doxorubicin Impurity)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.