missing translation for 'onlineSavingsMsg'
Learn More

6,8-Dibromoimidazo[1,2-a]pyrazine, 95%, Thermo Scientific Chemicals

Product Code. 15445378
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445378 5 g 5g
15435378 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445378 Supplier Thermo Scientific Alfa Aesar Supplier No. H59151.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63744-22-9 | Molecular Formula: C6H3Br2N3 | Molecular Weight: 276.92 g/mol

It is an Intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an Intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63744-22-9
Molecular Formula C6H3Br2N3
Molecular Weight (g/mol) 276.92
MDL Number MFCD08460056
InChI Key UQCZZGIPIMJBCL-UHFFFAOYSA-N
Synonym 6,8-dibromoimidazo 1,2-a pyrazine, imidazo 1,2-a pyrazine, 6,8-dibromo, 6,8-dibromo-imidazo 1,2-a pyrazine, 6,8-dibromoimidazol 1,2-a pyrazine, pubchem14656, acmc-1b8pv, ksc351o9d, buttpark 180\02-82, 6,8 dibromoimidazo 1,2-a pyrazine, 6,8-dibomoimidazo 1,2-a pyrazine
PubChem CID 15025843
IUPAC Name 6,8-dibromoimidazo[1,2-a]pyrazine
SMILES BrC1=CN2C=CN=C2C(Br)=N1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 276.92
Percent Purity 95%
Chemical Name or Material 6,8-Dibromoimidazo[1,2-a]pyrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.