missing translation for 'onlineSavingsMsg'
Learn More

6-Alpha-D-Glucopyranosyl Maltotriose-13C6, TRC

Product Code. 30004649
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
5 mg
Unit Size:
0.25mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004649 5 mg 5mg
30005924 1 mg 1mg
30010581 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004649 Supplier Toronto Research Chemicals Supplier No. TRCG4191035MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C1813C6H42O21
Molecular Weight (g/mol) 672.53
Synonym 4-O-[4-O-(6-O-α-D-Glucopyranosyl-α-D-glucopyranosyl)-α-D-glucopyranosyl]-D-glucopyranose-13C6, Glc-α-1,6-Glc-α-1,4-Glc-α-1,4Glc-13C6;
SMILES O[13C@H]1[13C@H](O)[13C@@H](O)[13C@@H](OC[C@@H]2[C@@H](O)[C@H](O)[C@@H](O)[C@@H](O[C@H]3[C@H](O)[C@@H](O)[C@@H](O[C@H]4[C@H](O)[C@@H](O)C(O)O[C@@H]4CO)O[C@@H]3CO)O2)O[13C@@H]1[13CH2]O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C24H42O21/c25-1-5-9(28)11(30)16(35)22(41-5)39-4-8-10(29)12(31)17(36)23(43-8)45-20-7(3-27)42-24(18(37)14(20)33)44-19-6(2-26)40-21(38)15(34)13(19)32/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10-,11+,12+,13-,14-,15-,16-,17-,18-,19-,20-,21?,22+,23-,24-/m1/s1/i1+1,5+1,9+1,11+1,16+1,22+1
Chemical Name or Material 6-alpha-D-Glucopyranosyl Maltotriose-13C6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.