missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-hexanol, 97%, Thermo Scientific Chemicals

Product Code. 11319015
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319015 1 g 1g
11329015 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319015 Supplier Thermo Scientific Alfa Aesar Supplier No. L14157.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 4048-33-3 | Molecular Formula: C6H15NO | Molecular Weight: 117.192 g/mol

6-Amino-1-hexanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-1-hexanol is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 4048-33-3
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00008241
InChI Key SUTWPJHCRAITLU-UHFFFAOYSA-N
Synonym 6-amino-1-hexanol, 1-hexanol, 6-amino, 6-aminohexanol, 1-amino-6-hexanol, hexanol, 6-amino, 6-hydroxyhexylamine, aminohexyl alcohol, amidohexylalkohol, amidohexylalkohol german, 6-amino-hexan-1-ol
PubChem CID 19960
IUPAC Name 6-aminohexan-1-ol
SMILES C(CCCO)CCN

Tekniske data

Melting Point 55°C to 58°C
Boiling Point 136°C to 139°C (30 mmHg)
Flash Point 133°C (271°F)
Quantity 1 g
Beilstein 1732524
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material 6-Amino-1-hexanol

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.