missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-chloro-5-nitropyridine, TRC

Product Code. 30316069
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316069 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316069 Supplier Toronto Research Chemicals Supplier No. TRCA6034952.5G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Amino-2-chloro-5-nitropyridine
CAS 27048-04-0
Molecular Formula C5H4ClN3O2
Quantity 2.5 g
Synonym 6-Chloro-3-nitro-2-pyridinamine, 2-Amino-6-chloro-3-nitro-pyridine, 6-Chloro-3-nitropyridin-2-ylamine
InChI Formula InChI=1S/C5H4ClN3O2/c6-4-2-1-3(9(10)11)5(7)8-4/h1-2H,(H2,7,8)
SMILES Nc1nc(Cl)ccc1[N+](=O)[O-]
IUPAC Name 6-chloro-3-nitropyridin-2-amine
Molecular Weight (g/mol) 173.56
Formula Weight 172.9992
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.