missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-methylnicotinonitrile, 97%, Maybridge

Product Code. 10289002
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10289002 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10289002 Supplier Thermo Scientific Maybridge Supplier No. MO01041DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Specifikationer

CAS 183428-90-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H7N3
MDL Number MFCD05865116
Quantity 1 g
Synonym 6-amino-2-methylnicotinonitrile, 6-amino-3-cyano-2-methylpyridine, 2-amino-5-cyano-6-methylpyridine, 3-pyridinecarbonitrile, 6-amino-2-methyl, 6-amino-2-methyl-nicotinonitrile, pubchem14519, acmc-20ahz0, 6-amino-2-methyl-pyridine-3-carbonitrile, 6-amino-2-methyl nicotinonitrile, 2-methyl-3-cyano-6-amino-pyridine
InChI Key OZAIJUZOQOBQDW-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=N1)N)C#N
IUPAC Name 6-amino-2-methylpyridine-3-carbonitrile
Molecular Weight (g/mol) 133.154
PubChem CID 2794662
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 6-Amino-2-methylnicotinonitrile
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.