missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-methylquinoline, 97%

Product Code. 11481718
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481718 1 g 1g
15463077 250 mg 250mg
11491718 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481718 Supplier Thermo Scientific Alfa Aesar Supplier No. A13967.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65079-19-8 | C10H10N2 | 158.204 g/mol

6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65079-19-8
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00052600
InChI Key TYJFYUVDUUACKX-UHFFFAOYSA-N
PubChem CID 103148
IUPAC Name 2-methylquinolin-6-amine
SMILES CC1=NC2=C(C=C1)C=C(C=C2)N

Specifications

Melting Point 189°C to 191°C
Quantity 1 g
Beilstein 114795
Synonym 6-amino-2-methylquinoline, 2-methyl-6-quinolinamine, 6-quinolinamine, 2-methyl, 6-aminoquinaldine, 2-methyl-6-aminoquinoline, 6-quinolinamine,2-methyl, 2-methyl-quinolin-6-ylamine, 2-methyl-6-quinolylamine, mqn, 2-methylquinoline-6-amine
Solubility Information Soluble in water.
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 6-Amino-2-methylquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.