missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-5-bromoquinoxaline, TRC

Product Code. 30134380
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134380 25 g 25g
17962095 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134380 Supplier Toronto Research Chemicals Supplier No. TRCA60188525G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50358-63-9
Molecular Formula C8 H6 Br N3
Molecular Weight (g/mol) 224.06
Synonym 5-Bromo-6-quinoxalinamine, 5-Bromo-6-aminoquinoxaline, 5-Bromoquinoxalin-6-amine, 6-Amino-5-bromoquinoxaline
IUPAC Name 5-bromoquinoxalin-6-amine
SMILES Nc1ccc2nccnc2c1Br

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H6BrN3/c9-7-5(10)1-2-6-8(7)12-4-3-11-6/h1-4H,10H2
Formula Weight 222.9745
Chemical Name or Material 6-Amino-5-bromoquinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.