missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-7-bromo-9-methyl-7-deazapurine, 97%, Thermo Scientific Chemicals

Product Code. 15456338
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456338 1 g 1g
15466338 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456338 Supplier Thermo Scientific Alfa Aesar Supplier No. H59897.03
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1337532-51-0 | Molecular Formula: C7H7BrN4 | Molecular Weight: 227.065 g/mol

It is used in the preparation of indoline derivatives as PERK inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of indoline derivatives as PERK inhibitors.

Solubility
Soluble in dichloromethane.

Notes
Store at room temperature. Keep away from oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1337532-51-0
Molecular Formula C7H7BrN4
Molecular Weight (g/mol) 227.065
MDL Number MFCD20486185
InChI Key SCHJLAFNBDGWJN-UHFFFAOYSA-N
Synonym 5-bromo-7-methyl-7h-pyrrolo 2,3-d pyrimidin-4-amine, 5-bromo-7-methylpyrrolo 2,3-d pyrimidin-4-amine, 6-amino-7-bromo-9-methyl-7-deazapurine, 5-bromo-7-methyl-7h-pyrrolo-2,3-d pyrimidin-4-amine, 7h-pyrrolo 2,3-d pyrimidin-4-amine,5-bromo-7-methyl, 7h-pyrrolo 2,3-d pyrimidin-4-amine, 5-bromo-7-methyl
PubChem CID 66560938
IUPAC Name 5-bromo-7-methylpyrrolo[2,3-d]pyrimidin-4-amine
SMILES CN1C=C(C2=C1N=CN=C2N)Br

Specifications

Quantity 1 g
Beilstein 21981671
Solubility Information Soluble in dichloromethane.
Formula Weight 227.06
Percent Purity 97%
Chemical Name or Material 6-Amino-7-bromo-9-methyl-7-deazapurine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.