missing translation for 'onlineSavingsMsg'
Learn More

6-Aminobenzothiazole, 98+%

Product Code. 11484324
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11484324 1 g 1g
11494324 5 g 5g
11404334 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11484324

Brand: Thermo Scientific Alfa Aesar A12601.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 533-30-2 | C7H6N2S | 150.199 g/mol

6-Aminobenzothiazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aminobenzothiazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-30-2
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.199
MDL Number MFCD00015461
InChI Key FAYAYUOZWYJNBD-UHFFFAOYSA-N
Synonym 6-aminobenzothiazole, 6-benzothiazolamine, benzo d thiazol-6-amine, 6-amino-benzothiazole, benzothiazole, 6-amino, benzothiazol-6-amine, benzothiazol-6-ylamine, unii-9yfg4wuj1g, 9yfg4wuj1g, 1,3-benzothiazol-6-ylamine
PubChem CID 68288
IUPAC Name 1,3-benzothiazol-6-amine
SMILES C1=CC2=C(C=C1N)SC=N2

Specifications

Melting Point 86°C to 90°C
Quantity 1 g
Beilstein 116381
Merck Index 14,425
Solubility Information Slightly soluble in water.
Formula Weight 150.2
Percent Purity ≥98%
Chemical Name or Material 6-Aminobenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.