missing translation for 'onlineSavingsMsg'
Learn More

6-Aminobenzoxazole, 95%, Thermo Scientific™

Product Code. 15487437
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487437 250 mg 250mg
15467437 1 g 1g
15477437 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15487437 Supplier Thermo Scientific Alfa Aesar Supplier No. H34485.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
6-Aminobenzoxazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177492-52-3
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD03791187
InChI Key ZJYIRVSPPOOPCL-UHFFFAOYSA-N
Synonym benzo d oxazol-6-amine, 6-aminobenzoxazole, 6-benzoxazolamine, benzooxazol-6-ylamine, 6-amino-benzoxazole, benzoxazole-6-ylamine, pubchem17704, 6-amino-1,3-benzoxazole, #
PubChem CID 584120
IUPAC Name 1,3-benzoxazol-6-amine
SMILES C1=CC2=C(C=C1N)OC=N2

Specifications

Melting Point 95°C to 97°C
Color Orange to Brown
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 134.1
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 6-Aminobenzoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.