missing translation for 'onlineSavingsMsg'
Learn More

6-Aminohexanoic Acid, TRC

Product Code. 30325720
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325720 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325720 Supplier Toronto Research Chemicals Supplier No. TRCA60998510MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aminocaproic acid
CAS 60-32-2
Molecular Formula C6 H13 N O2
Quantity 10 mg
Synonym Aminocaproic Acid, ε-Aminocaproic acid, ε-Amino-n-hexanoic acid, ε-Aminocaproic acid, ε-Aminohexanoic acid, ε-Leucine, ε-Norleucine, ω-Aminocaproic acid, ω-Aminohexanoic acid, 177 J.D., 6-Amino-n-hexanoic acid, 6-Aminocaproic acid, 6-Aminohexanoic acid, ACS, Acepramin, Acepramine, Acikaprin, Afibrin, Amicar, Amikar, Aminokapron, CL 10304, CY 116, Caplamin, Capramol, Caprocid, Caprolisin, EACA, EACS, Epsamon, Epsicapron, Epsikapron, Epsilcapramin, Epsilon S, Hemocaprol, Hemopar, Hepin, Ipsilon, NSC 212532, NSC 26154, NSC 400230, Respramin
InChI Formula InChI=1S/C6H13NO2/c7-5-3-1-2-4-6(8)9/h1-5,7H2,(H,8,9)
SMILES NCCCCCC(=O)O
IUPAC Name 6-aminohexanoic acid
Molecular Weight (g/mol) 131.17
Formula Weight 131.0946
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.