missing translation for 'onlineSavingsMsg'
Learn More

6-(Aminomethyl)indole, 97%

Product Code. 15450878
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450878 250 mg 250mg
15430878 1 g 1g
15440878 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450878 Supplier Thermo Scientific Alfa Aesar Supplier No. H54640.MD
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3468-17-5 | C9H10N2 | 146.19 g/mol

6-(Aminomethyl)indole is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(Aminomethyl)indole is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3468-17-5
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD06213839
InChI Key FURRUNQWZZOXOT-UHFFFAOYSA-N
Synonym 1h-indole-6-methanamine, 1h-indol-6-yl methanamine, 1h-indol-6-ylmethylamine, 1-1h-indol-6-yl methanamine, 6-aminomethylindole, 6-aminomethyl indole, 1h-indol-6-ylmethyl amine, indole, 6-aminomethyl, c-1h-indol-6-yl-methylamine
PubChem CID 17221101
IUPAC Name 1H-indol-6-ylmethanamine
SMILES NCC1=CC=C2C=CNC2=C1

Specifications

Quantity 250 mg
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 6-(Aminomethyl)indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.