missing translation for 'onlineSavingsMsg'
Learn More

6-Aminonicotinamide, 98%

Product Code. 15214604 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15214604 5 g Glass bottle 5g
15204604 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15214604 Supplier Thermo Scientific Acros Supplier No. 104150050
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 329-89-5 | C6H7N3O | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-89-5
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006327
InChI Key ZLWYEPMDOUQDBW-UHFFFAOYSA-N
Synonym 6-aminonicotinamide, 2-amino-5-carbamoylpyridine, 3-pyridinecarboxamide, 6-amino, 6-amino-3-pyridinecarboxamide, 6-aminonicotinic acid amide, 6-aminonikotinsaeureamid, nicotinamide, 6-amino, aminonicotinamide, 6-an, 6-amino-nicotinsaeureamid
PubChem CID 9500
ChEBI CHEBI:74514
IUPAC Name 6-aminopyridine-3-carboxamide
SMILES NC(=O)C1=CC=C(N)N=C1

Specifications

Melting Point 245°C to 248°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2N(C5H3N)CONH2
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 137.4
Percent Purity 98%
Chemical Name or Material 6-Aminonicotinamide
compliance-icons
Product Identifier
  • 6-Aminonicotinamide
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.