missing translation for 'onlineSavingsMsg'
Learn More

6-Aminophthalide, 95%

Product Code. 11403148
Change view
Click to view available options
Quantity:
0.5 g
2.5 g
10 g
Unit Size:
0.5g
10g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403148 0.5 g 0.5g
11483138 2.5 g 2.5g
11493138 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403148 Supplier Thermo Scientific Alfa Aesar Supplier No. A18896.02
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 57319-65-0 | C8H7NO2 | 149.149 g/mol

It is used to produce 6-acetylamino-phthalide. It is also used as a starting material in the reduction reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce 6-acetylamino-phthalide. It is also used as a starting material in the reduction reaction.

Solubility
Soluble in water 1360 mg/L 25°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57319-65-0
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00033530
InChI Key ZIJZDNKZJZUROE-UHFFFAOYSA-N
Synonym 6-aminophthalide, 6-aminoisobenzofuran-1 3h-one, 6-amino-1,3-dihydroisobenzofuran-1-one, 6-amino-2-benzofuran-1 3h-one, 6-amino-3h-isobenzofuran-1-one, 1 3h-isobenzofuranone, 6-amino, 6-amino-1,3-dihydro-2-benzofuran-1-one, 6-amino-1 3h-isobenzofuranone, 6-amino-3-hydroisobenzofuran-1-one, 6-aminophthalid
PubChem CID 93631
IUPAC Name 6-amino-3H-2-benzofuran-1-one
SMILES C1C2=C(C=C(C=C2)N)C(=O)O1

Specifications

Melting Point 182°C to 185°C
Quantity 0.5 g
Solubility Information Soluble in water 1360mg/L 25°C.
Formula Weight 149.15
Percent Purity 95%
Chemical Name or Material 6-Aminophthalide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.