missing translation for 'onlineSavingsMsg'
Learn More

6-Aza-2-thiothymine, 98%

Product Code. 11492207
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492207 25 g 25g
11482207 5 g 5g
11402217 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11492207 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A14167.14
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 615-76-9 | C4H5N3OS | 143.164 g/mol

6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

Solubility
Soluble in water (partly), and formic acid (5 %).

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-76-9
Molecular Formula C4H5N3OS
Molecular Weight (g/mol) 143.164
MDL Number MFCD00006458
InChI Key NKOPQOSBROLOFP-UHFFFAOYSA-N
Synonym 6-aza-2-thiothymine, 2-thio-6-azathymine, 6-azathiothymine, 6-methyl-3-thioxo-3,4-dihydro-1,2,4-triazin-5 2h-one, 3-mercapto-6-methyl-1,2,4-triazin-5 2h-one, 1,2,4-triazin-5 2h-one, 3,4-dihydro-6-methyl-3-thioxo, 3-mercapto-6-methyl-as-triazin-5-ol, as-triazin-5-ol, 3-mercapto-6-methyl, 6-methyl-3-sulfanyl-1,2,4-triazin-5-ol, 3,4-dihydro-6-methyl-3-thioxo-1,2,4-triazin-5 2h-one
PubChem CID 1263666
IUPAC Name 6-methyl-3-sulfanylidene-2H-1,2,4-triazin-5-one
SMILES CC1=NNC(=S)NC1=O

Specifications

Melting Point 218°C to 221°C
Quantity 25 g
Beilstein 4861
Solubility Information Soluble in water (partly),and formic acid (5 %).
Formula Weight 143.17
Percent Purity 98%
Chemical Name or Material 6-Aza-2-thiothymine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.