missing translation for 'onlineSavingsMsg'
Learn More

6-Benzylaminopurine, MP Biomedicals™

Product Code. 11334129 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334129 25 g 25g
11324129 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334129 Supplier MP Biomedicals Supplier No. 0210091225
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1214-39-7
Molecular Formula C12H11N5
Molecular Weight (g/mol) 225.26
MDL Number MFCD00005572
InChI Key NWBJYWHLCVSVIJ-UHFFFAOYSA-N
Synonym 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
PubChem CID 62389
ChEBI CHEBI:29022
IUPAC Name N-benzyl-7H-purin-6-amine
SMILES C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1

Specifications

Quantity 25 g
Formula Weight 225.3
Physical Form Solid
Chemical Name or Material 6-Benzylaminopurine
compliance-icons
Product Identifier
  • 6-Benzylaminopurine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 6-Benzylaminopurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.