missing translation for 'onlineSavingsMsg'
Learn More

6-Benzylaminopurine, MP Biomedicals

Product Code. 11314129 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314129 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11314129 Supplier MP Biomedicals Supplier No. 02100912.5
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

  • 6-Benzylaminopurine, benzyl adenine (BAP) is a synthetic cytokinin which together with auxins elicits plant growth and development responses
  • BAP is a widely use cytokinin supplement to plant growth media such as Murashige and Skoog medium, Gamborg’s medium, and Chu’s N6 medium
  • Inhibitor of respiratory kinase in plants
  • Increases post-harvest life of green vegetables

TRUSTED_SUSTAINABILITY

Specifications

CAS 1214-39-7
Melting Point 226°C to 236°C
Color White
Molecular Formula C12H11N5
Linear Formula C12H11N5
MDL Number MFCD00005572
Quantity 500 mg
Synonym 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
Solubility Information Soluble in glacial Acetic Acid (50mg/mL). Soluble in concentrated ammonium hydroxide, 0.1 N Sodium hydroxide (10mg/mL - slightly hazy and yellowish solution), methanol, or dioxane.
InChI Key NWBJYWHLCVSVIJ-UHFFFAOYSA-N
SMILES C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1
IUPAC Name N-benzyl-7H-purin-6-amine
Molecular Weight (g/mol) 225.26
PubChem CID 62389
ChEBI CHEBI:29022
Formula Weight 225.25
Physical Form Powder
Chemical Name or Material 6-BENZYLAMINOPURINE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.