missing translation for 'onlineSavingsMsg'
Learn More

6-Benzylaminopurine, TRC

Product Code. 30135738
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135738 50 g 50g
30138745 100 g 100g
30008094 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135738 Supplier Toronto Research Chemicals Supplier No. TRCB22630550G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214-39-7
Molecular Formula C12 H11 N5
Molecular Weight (g/mol) 225.25
Synonym 1H-Purin-6-amine, N-(phenylmethyl)- (9CI), Adenine, N-benzyl- (7CI,8CI), Adenine, N6-benzyl- (6CI), N-(Phenylmethyl)-9H-purin-6-amine, 6-(Benzylamino)purine, 6-(N-Benzylamino)purine, 6-BA, 6-BAP, 6-Benzyladenine, 6-Benzylaminoadenine, ABG 3034, BA, BA (growth stimulant), BAP, BAP (growth stimulant), Benzyladenine, Benzylaminopurine, Bongrow, CTP, CyLex, Cytokinin B, MaxCel, N-(Phenylmethyl)-1H-purin-6-amine, N-Benzyladenine, N6-(Benzylamino)purine, N6-Benzyladenine, N6-Benzylaminopurine, NSC 40818, Paturyl, Paturyl 10WSC, Pro-Shear, SD 4901, SQ 4609, TOG-L 101
IUPAC Name N-benzyl-7H-purin-6-amine
SMILES C(Nc1ncnc2nc[nH]c12)c3ccccc3

Specifications

Quantity 50 g
InChI Formula InChI=1S/C12H11N5/c1-2-4-9(5-3-1)6-13-11-10-12(15-7-14-10)17-8-16-11/h1-5,7-8H,6H2,(H2,13,14,15,16,17)
Formula Weight 225.1014
Chemical Name or Material 6-Benzylaminopurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.