missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyloxy-1-BOC-indole-2-boronic acid, 97%, Thermo Scientific™

Product Code. 10658429 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10658429 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10658429 Supplier Thermo Scientific Acros Supplier No. 430330010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 850568-66-0
CAS Min % 96.0
CAS Max % 100.0
Melting Point 102.0°C to 106.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C20H22BNO5
MDL Number MFCD06659831
Quantity 1 g
Synonym 6-benzyloxy-1-boc-indole-2-boronic acid, 6-benzyloxy-1-tert-butoxycarbonyl-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-6-benzyloxy-1h-indol-2-yl-2-boronic acid, 6-benzyloxy-1h-indole-2-boronic acid, n-boc protected, 6-benzyloxy-1-tert-butoxycarbonyl indol-2-ylboronic acid, 1h-indole-1-carboxylicacid, 2-borono-6-phenylmethoxy-, 1,1-dimethylethyl ester, acmc-209q2h, 6-benzyloxy-1-boc-2-indoleboronic acid, 6-benzyloxyindole-2-boronic acid, n-boc protected
InChI Key OGONSJPAZMVGTG-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1C(=CC2=CC=C(OCC3=CC=CC=C3)C=C12)B(O)O
IUPAC Name [1-[(2-methylpropan-2-yl)oxycarbonyl]-6-phenylmethoxyindol-2-yl]boronic acid
Molecular Weight (g/mol) 367.21
PubChem CID 22309458
Formula Weight 367.21
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-Benzyloxy-1-BOC-indole-2-boronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 6-Benzyloxy-1-Boc-indole-2-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.