missing translation for 'onlineSavingsMsg'
Learn More

6-Boc-1-oxa-6-azaspiro[2.5]octane, 97%

Product Code. 15497057
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497057 1 g 1g
15417067 250 mg 250mg
15407067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15497057 Supplier Thermo Scientific Alfa Aesar Supplier No. H34134.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 147804-30-6 | C11H19NO3 | 213.277 g/mol

It is used as a pharmaceutical intermediate. It is also used as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. It is also used as an intermediate in organic syntheses.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and place it in a cool, dry and well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147804-30-6
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.277
MDL Number MFCD07779385
InChI Key ULSBMKGFFFMGOI-UHFFFAOYSA-N
Synonym tert-butyl 1-oxa-6-azaspiro 2.5 octane-6-carboxylate, 1-oxa-6-azaspiro 2.5 octane-6-carboxylic acid, 1,1-dimethylethyl ester, 6-boc-1-oxa-6-azaspiro 2.5 octane, tert-butyl 1-oxa-6-azaspiro 2,5 octane-6-carboxylate, n-boc-1-oxa-6-azaspiro 2.5 octane, 6-n-boc-1-oxa-6-azaspiro 2.5 octane, 6-tert-butoxycarbonyl-1-oxa-6-azaspiro 2.5 octane, tert-butyl 2-oxa-6-azaspiro 2.5 octane-6-carboxylate, 1-oxa-6-aza-spiro 2.5 octane-6-carboxylic acid tert-butyl ester, tert-butyl 6-azaspiro 2.5 octane-1-oxa-6-carboxylate
PubChem CID 22135564
IUPAC Name tert-butyl 1-oxa-6-azaspiro[2.5]octane-6-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2(CC1)CO2

Specifications

Melting Point 56°C to 58°C
Quantity 1 g
Formula Weight 213.28
Percent Purity 97%
Chemical Name or Material 6-Boc-1-oxa-6-azaspiro[2.5]octane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.