missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,3-benzothiazole, 97%, Thermo Scientific™

Product Code. 10666614
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10666614 5 g Amber Glass Bottle 5g
10279712 250 mg Amber Glass Bottle 250mg
10416082 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10666614 Supplier Thermo Scientific Maybridge Supplier No. CC05910DE
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 53218-26-1
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD04115372
InChI Key YJOUISWKEOXIMC-UHFFFAOYSA-N
Synonym 6-bromobenzothiazole, 6-bromobenzo d thiazole, benzothiazole, 6-bromo, 6-bromo-benzothiazole, 6-bromo-benzo d thiazole, pubchem21861, acmc-209l4t, 6-bromanyl-1,3-benzothiazole, benzothiazole, 6-bromo-9ci
PubChem CID 2795171
IUPAC Name 6-bromo-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Br)SC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 214.09
Percent Purity 97%
Chemical Name or Material 6-Bromo-1, 3-benzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.