missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,4-benzodioxane, 98%

Product Code. 11481647
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481647 5 g 5g
11471647 1 g 1g
15433297 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481647 Supplier Thermo Scientific Alfa Aesar Supplier No. B25625.06
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 52287-51-1 | C8H7BrO2 | 215.046 g/mol

6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52287-51-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00040750
InChI Key LFCURAJBHDNUNG-UHFFFAOYSA-N
Synonym 6-bromo-1,4-benzodioxane, 6-bromo-2,3-dihydrobenzo b 1,4 dioxine, 3,4-ethylenedioxybromobenzene, 6-bromo-2,3-dihydro-1,4-benzodioxin, 1,4-benzodioxin, 6-bromo-2,3-dihydro, 4-bromo-1,2-ethylenedioxybenzene, 3,4-ethylendioxybromobenzene, 6-bromo-2,3-dihydro-benzo 1,4 dioxine, pubchem3736, acmc-209kyk
PubChem CID 104141
IUPAC Name 6-bromo-2,3-dihydro-1,4-benzodioxine
SMILES C1COC2=C(O1)C=CC(=C2)Br

Specifications

Density 1.598
Boiling Point 259°C to 260°C
Flash Point >110°C (230°F)
Refractive Index 1.589
Quantity 5 g
Beilstein 6223
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 6-Bromo-1,4-benzodioxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.