missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2,4-dichloroquinazoline, 97%

Product Code. 15490379
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490379 1 g 1g
15410389 250 mg 250mg
15400389 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490379 Supplier Thermo Scientific Alfa Aesar Supplier No. H63603.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102393-82-8 | C8H3BrCl2N2 | 277.93 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102393-82-8
Molecular Formula C8H3BrCl2N2
Molecular Weight (g/mol) 277.93
MDL Number MFCD09744007
InChI Key LBAYOWRVZAKPLS-UHFFFAOYSA-N
PubChem CID 10107568
IUPAC Name 6-bromo-2,4-dichloroquinazoline
SMILES C1=CC2=C(C=C1Br)C(=NC(=N2)Cl)Cl

Specifications

Color White
Quantity 1 g
Synonym quinazoline, 6-bromo-2,4-dichloro, 2,4-dichloro-6-bromoquinazoline, 6-bromo-2,4-dichloro-quinazoline, quinazoline,6-bromo-2,4-dichloro, pubchem14690, acmc-1c6nj, abbypharma ap-16-10560, 6-bromanyl-2,4-bis chloranyl quinazoline
Solubility Information Slightly soluble in water.
Formula Weight 277.93
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Bromo-2,4-dichloroquinazoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.