missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-chlorobenzothiazole, 97%

Product Code. 15464607
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15464607 1 g 1g
15474607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464607 Supplier Thermo Scientific Alfa Aesar Supplier No. H31962.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 80945-86-4 | C7H3BrClNS | 248.522 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80945-86-4
Molecular Formula C7H3BrClNS
Molecular Weight (g/mol) 248.522
MDL Number MFCD04971822
InChI Key IJQSMNIZBBEBKI-UHFFFAOYSA-N
PubChem CID 2049871
IUPAC Name 6-bromo-2-chloro-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Br)SC(=N2)Cl

Specifications

Melting Point 98°C to 100°C
Quantity 1 g
Synonym 6-bromo-2-chlorobenzothiazole, 6-bromo-2-chlorobenzo d thiazole, 6-bromo-2-chloro-benzothiazole, 2-chloro-6-bromobenzothiazole, benzothiazole, 6-bromo-2-chloro, 6-bromo-2-chlorbenzothiazole, benzothiazole,6-bromo-2-chloro, zlchem 563, pubchem11132, acmc-1bkg3
Formula Weight 248.53
Percent Purity 97%
Chemical Name or Material 6-Bromo-2-chlorobenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.