missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2H-1,4-benzoxazin-3(4H)-one, 95%

Product Code. 11471730
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471730 10 g 10g
11461730 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471730 Supplier Thermo Scientific Alfa Aesar Supplier No. H27493.09
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24036-52-0 | C8H6BrNO2 | 228.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24036-52-0
Molecular Formula C8H6BrNO2
Molecular Weight (g/mol) 228.045
MDL Number MFCD00461173
InChI Key UQCFMEFQBSYDHY-UHFFFAOYSA-N
Synonym 6-bromo-2h-1,4-benzoxazin-3 4h-one, 6-bromo-2h-benzo b 1,4 oxazin-3 4h-one, 6-bromo-3-oxo-4h-benzo 1,4 oxazine, 6-bromo-1,4-benzoxazin-3-one, 6-bromo-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2h-1,4-benzoxazin-3 4h-one, 6-bromo, 6-bromo-2,4-dihydro-1,4-benzoxazin-3-one, 6-bromo-4h-benzo 1,4 oxazin-3-one, pubchem16746
PubChem CID 16218142
IUPAC Name 6-bromo-4H-1,4-benzoxazin-3-one
SMILES C1C(=O)NC2=C(O1)C=CC(=C2)Br

Specifications

Melting Point 220°C to 225°C
Quantity 10 g
Formula Weight 228.04
Percent Purity 95%
Chemical Name or Material 6-Bromo-2H-1,4-benzoxazin-3(4H)-one

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.