missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-pyridazinamine, 97%, Thermo Scientific™

Product Code. 10655155
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10655155 5 g Amber Glass Bottle 5g
10390453 250 mg Amber Glass Bottle 250mg
10630105 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10655155 Supplier Thermo Scientific Maybridge Supplier No. MO07086DE
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 88497-27-2
Molecular Formula C4H4BrN3
Molecular Weight (g/mol) 174.001
MDL Number MFCD01646092
InChI Key FXTDHDQFLZNYKW-UHFFFAOYSA-N
PubChem CID 2794779
IUPAC Name 6-bromopyridazin-3-amine
SMILES C1=CC(=NN=C1N)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 3-amino-6-bromopyridazine, 6-bromo-3-pyridazinamine, 6-bromo-pyridazin-3-ylamine, 2-amino-5-bromopyridazine, 3-pyridazinamine,6-bromo, 3-pyridazinamine, 6-bromo, 6-bromopyridazine-3-ylamine, 6-bromo-3-pyridazineamine, 3-bromo-6-aminopyridazine, acmc-20a0gf
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 6-Bromo-3-pyridazinamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.