missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4-methyl-3-nitroquinoline, 96%, Thermo Scientific Chemicals

Product Code. 15400978
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400978 1 g 1g
15410978 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400978 Supplier Thermo Scientific Alfa Aesar Supplier No. H54769.03
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1185310-49-9 | Molecular Formula: C10H7BrN2O2 | Molecular Weight: 267.082 g/mol

6-Bromo-4-methyl-3-nitroquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-4-methyl-3-nitroquinoline is used as pharmaceutical intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185310-49-9
Molecular Formula C10H7BrN2O2
Molecular Weight (g/mol) 267.082
MDL Number MFCD12761249
InChI Key MYJZQOHLSJFXDS-UHFFFAOYSA-N
PubChem CID 45789224
IUPAC Name 6-bromo-4-methyl-3-nitroquinoline
SMILES CC1=C2C=C(C=CC2=NC=C1[N+](=O)[O-])Br

Specifications

Melting Point 152°C to 155°C
Quantity 1 g
Beilstein 19927450
Formula Weight 267.08
Percent Purity 96%
Chemical Name or Material 6-Bromo-4-methyl-3-nitroquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.