missing translation for 'onlineSavingsMsg'
Learn More

6-Bromochroman, 97%, Thermo Scientific™

Product Code. 10041534
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041534 1 g Amber Glass Bottle 1g
10396614 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041534 Supplier Thermo Scientific Maybridge Supplier No. CC32810DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3875-78-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD10698725
InChI Key KPFDABVKWKOIME-UHFFFAOYSA-N
Synonym 6-bromochroman, 6-bromo-chroman, 6-bromochromane, 6-bromo-3,4-dihydro-2h-1-benzopyran, 2h-1-benzopyran,6-bromo-3,4-dihydro, 6-bromanyl-3,4-dihydro-2h-chromene, 2h-1-benzopyran, 6-bromo-3,4-dihydro
PubChem CID 10856814
IUPAC Name 6-bromo-3,4-dihydro-2H-chromene
SMILES C1CC2=C(C=CC(=C2)Br)OC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 213.07
Percent Purity 97%
Chemical Name or Material 6-Bromochroman
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.