missing translation for 'onlineSavingsMsg'
Learn More

6-Bromohexene, TRC

Product Code. 30330757
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330757 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330757 Supplier Toronto Research Chemicals Supplier No. TRCB6819305G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Bromohexene
CAS 2695-47-8
Molecular Formula C6H11Br
Quantity 5 g
Synonym 1-Bromo-5-hexene, 5-Hexen-1-yl Bromide, 5-Hexenyl Bromide, 6-Bromo-1-hexene
InChI Formula InChI=1S/C6H11Br/c1-2-3-4-5-6-7/h2H,1,3-6H2
SMILES BrCCCCC=C
IUPAC Name 6-bromohex-1-ene
Molecular Weight (g/mol) 163.06
Formula Weight 162.0044
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.