missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindole-2-carboxylic acid, 97%

Product Code. 15450689
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450689 250 mg 250mg
15430689 1 g 1g
15440689 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450689 Supplier Thermo Scientific Alfa Aesar Supplier No. H63799.MD
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16732-65-3 | C9H6BrNO2 | 240.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16732-65-3
Molecular Formula C9H6BrNO2
Molecular Weight (g/mol) 240.056
MDL Number MFCD02664469
InChI Key SVBVYRYROZWKNJ-UHFFFAOYSA-N
Synonym 6-bromoindole-2-carboxylic acid, 6-bromo-2-indolecarboxylic acid, 1h-indole-2-carboxylic acid, 6-bromo, 6-bromoindole-2-carboxylicacid, zlchem 475, pubchem1664, acmc-1bqkh, intermediates-zcf02255, evoblocks eb21020, timtec-bb sbb014280
PubChem CID 4011696
IUPAC Name 6-bromo-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1Br)NC(=C2)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 240.05
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Bromoindole-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.