missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindole, 96%

Product Code. 10785905 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10785905 5 g Glass bottle 5g
10224774 250 mg Glass bottle 250mg
10602835 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10785905 Supplier Thermo Scientific Acros Supplier No. 397360050
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52415-29-9 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52415-29-9
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00238550
InChI Key MAWGHOPSCKCTPA-UHFFFAOYSA-N
Synonym 6-bromoindole, 1h-indole, 6-bromo, bromoindole-6, 6-bromo-indole, 6-brom-1h-indole, pubchem1662, 6-bromo-1-h-indole, 6-bromindol, maybridge1_006369
PubChem CID 676493
IUPAC Name 6-bromo-1H-indole
SMILES C1=CC(=CC2=C1C=CN2)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 92.0°C to 96.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 196.05
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 6-Bromoindole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.