missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoisatin, 95%

Product Code. 15395008 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15395008 5 g 5g
15305018 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15395008 Supplier Thermo Scientific Acros Supplier No. 455510050
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6326-79-0 | C8H4BrNO2 | 226.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6326-79-0
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.03
MDL Number MFCD01631138
InChI Key HVPQMLZLINVIHW-UHFFFAOYSA-N
Synonym 6-bromoisatin, 6-bromoindoline-2,3-dione, 6-bromo-isatin, 1h-indole-2,3-dione, 6-bromo, 6-bromoindole-2,3-dione, 6-bromo-2,3-dihydro-1h-indole-2,3-dione, 6-bromo-1h-benzo d azolidine-2,3-dione, 6-bromoindolin-2,3-dione, 6-bromo-2,3-indolinedione
PubChem CID 95716
SMILES BrC1=CC=C2C(NC(=O)C2=O)=C1

Specifications

Melting Point 274.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
IUPAC Name 6-bromo-2,3-dihydro-1H-indole-2,3-dione
Formula Weight 226.03
Percent Purity 95%
Chemical Name or Material 6-Bromoisatin
compliance-icons
Product Identifier
  • 6-Bromoisatin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.