missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoisoquinoline, 97%

Product Code. p-4228454 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10594208

Brand: Thermo Scientific Acros 429460010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34784-05-9 | C9H6BrN | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34784-05-9
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD04973299
InChI Key ZTEATMVVGQUULZ-UHFFFAOYSA-N
Synonym 6-bromo-isoquinoline, isoquinoline, 6-bromo, 6-bromo isoquinoline, zlchem 769, 6-bromoisoquinoline, pubchem17031, acmc-209ia9, ksc201e8h, monohydrate 1g
PubChem CID 313681
IUPAC Name 6-bromoisoquinoline
SMILES C1=CC2=C(C=CN=C2)C=C1Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 42.6°C to 43.6°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 208.06
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoisoquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 6-Bromoisoquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.