missing translation for 'onlineSavingsMsg'
Learn More

6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline, ≥95%, Thermo Scientific™

Product Code. 10314632
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10314632 10 g Amber Glass Bottle 10g
10121222 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10314632 Supplier Thermo Scientific Maybridge Supplier No. KM07330EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 123637-51-4
Molecular Formula C11H6BrClF3N
Molecular Weight (g/mol) 324.525
MDL Number MFCD00153079
InChI Key SJBKLFYWXYFAGT-UHFFFAOYSA-N
PubChem CID 2736346
IUPAC Name 6-(bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
SMILES C1=CC2=C(C=C1CBr)C(=CC(=N2)C(F)(F)F)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 6-bromomethyl-4-chloro-2-trifluoromethyl quinoline, quinoline,6-bromomethyl-4-chloro-2-trifluoromethyl, quinoline, 6-bromomethyl-4-chloro-2-trifluoromethyl, pubchem6004, acmc-1bzc1, buttpark 29\04-10, 6-bromomethyl-4-chloro-2-trifluoromethyl-quinoline, 2-trifluoromethyl-4-chloro-6-bromomethyl quinoline
Formula Weight 324.53
Percent Purity 95%
Chemical Name or Material 6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.