missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopiperonal, 98%

Product Code. 11478117
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478117 5 g 5g
11488117 25 g 25g
11498117 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478117 Supplier Thermo Scientific Alfa Aesar Supplier No. A17510.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15930-53-7 | C8H5BrO3 | 229.03 g/mol

6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.

Solubility
Soluble in DMSO, methanol

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15930-53-7
Molecular Formula C8H5BrO3
Molecular Weight (g/mol) 229.03
MDL Number MFCD00022952
InChI Key CSQUXTSIDQURDV-UHFFFAOYSA-N
Synonym 6-bromopiperonal, 6-bromo-benzo 1,3 dioxole-5-carbaldehyde, 2-bromo-4,5-methylenedioxybenzaldehyde, piperonal, 6-bromo, 6-bromobenzo d 1,3 dioxole-5-carbaldehyde, 6-bromo-1,3-benzodioxole-5-carboxaldehyde, 6-bromo-2h-1,3-benzodioxole-5-carbaldehyde, 1,3-benzodioxole-5-carboxaldehyde, 6-bromo, 6-bromobenzo 1,3 dioxole-5-carbaldehyde, benzaldehyde, 2-bromo-4,5-methylenedioxy
PubChem CID 95062
SMILES BrC1=CC2=C(OCO2)C=C1C=O

Specifications

Melting Point 127°C to 130°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in DMSO,methanol
IUPAC Name 6-bromo-1,3-benzodioxole-5-carbaldehyde
Formula Weight 229.03
Percent Purity 98%
Chemical Name or Material 6-Bromopiperonal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.